Isobutyraldehyde ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
I0101-25ML I0101-500ML
TCIAI0101-25MLEA 17.6 EUR
TCIAI0101-25ML TCIAI0101-500ML
Isobutyraldehyde ≥98.0% (by GC)
Isobutyraldehyde
Formule: C₄H₈O
Kookpunt: 65°C
Smeltpunt: -65°C
Dichtheid: 0,794 g/cm³ (20 °C)
Vlampunt: -25°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00006980
CAS nummer: 78-84-2
EINECS: 201-149-6
UN: 2045
ADR: 3,II
Merck Index: 14,05154
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a72b17243848bfb282438206002ebf0e5e5605aa&vid=c0ba3dca23c80f584ff2319e4de9f138f52806ae&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %