Methylisopropylketon ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

M0173-500ML M0173-25ML
TCIAM0173-500MLEA 50.6 EUR
TCIAM0173-500ML TCIAM0173-25ML
Methylisopropylketon ≥99.0% (by GC)
Methylisopropylketon
Formule: C₅H₁₀O
Kookpunt: 93°C
Smeltpunt: –92 °C
Dichtheid: 0,8046 g/cm³ (20 °C)
Vlampunt: 1°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008919
CAS nummer: 563-80-4
EINECS: 209-264-3
UN: 2397
ADR: 3,II
Merck Index: 14,06090
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18594a1b6a256278082a061f68ff117faf045cd3&vid=82dadf2236a837df9b34ec3ccba504630ebd3a2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR