5-Methyl-2-hexaanon ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Isoamyl methyl ketone, Methyl isoamyl ketone
I0087-25ML I0087-500ML
TCIAI0087-25MLEA 18.7 EUR
TCIAI0087-25ML TCIAI0087-500ML
5-Methyl-2-hexaanon ≥98.0% (by GC)
5-Methyl-2-hexaanon
Formule: C₇H₁₄O
MW: 114.19 g/mol
Kookpunt: 144°C
Smeltpunt: –74°C
Vlampunt: 36°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008950
CAS nummer: 110-12-3
EINECS: 203-737-8
UN: 2302
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cf0b6d70c679f5901d6cc74ff38e3e9889ad641&vid=478a9be903db5124bc507fb2a29f65e402327706&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %