1-Chloro-3-methylbutane ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
C0199-25ML C0199-500ML
TCIAC0199-25MLEA 38.5 EUR
TCIAC0199-25ML TCIAC0199-500ML
1-Chloro-3-methylbutane ≥98.0% (by GC)
1-Chloro-3-methylbutane
Formule: C₅H₁₁Cl
Kookpunt: 100°C
Dichtheid: 0,893 g/cm³ (20 °C)
Vlampunt: 16°C
MDL Nummer: MFCD00013693
CAS nummer: 107-84-6
EINECS: 203-525-5
UN: 1107
ADR: 3,II
Merck Index: 14,05116
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b52729e9963a7aee4fe67e1fac87807abb82e91&vid=d4ed7be4b00a62e0e96524c2cfdedf14cf282549&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %