2,2,4,4,6,8,8-Heptamethylnonane ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
H0365-25ML
H0365-500ML
H0365-100ML
TCIAH0365-25MLEA
34.1
EUR
TCIAH0365-25ML
TCIAH0365-500ML
TCIAH0365-100ML
2,2,4,4,6,8,8-Heptamethylnonane ≥97.0% (by GC)
2,2,4,4,6,8,8-Heptamethylnonane
Formule:
C₁₆H₃₄ Kookpunt: 247°C Vlampunt: 102°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008856 CAS nummer: 4390-04-9 EINECS: 224-506-8 ADR: 3,III |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...