2,2,4-Trimethylpentane 99,0 GC_ASSAY_METHOD
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
T0481-25ML T0481-500ML
TCIAT0481-25MLEA 17.6 EUR
TCIAT0481-25ML TCIAT0481-500ML
2,2,4-Trimethylpentane 99,0 GC_ASSAY_METHOD
2,2,4-Trimethylpentane
Formule: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Kookpunt: 99°C
Smeltpunt: -107
Vlampunt: –9°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008943
CAS nummer: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3 II
Merck Index: 14,05193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63ca50e132135ddafc11e06ca7ddd7b94f9cd90f&vid=c2527c68f68c98165f8ddb529daca68bcc71f3a5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless clear liquid
Purity (GC) min. 99.0 %