6-Bromo-1-hexene ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B2106-25G B2106-5G
TCIAB2106-25GEA 357 EUR
TCIAB2106-25G TCIAB2106-5G
6-Bromo-1-hexene ≥95.0% (by GC)
6-Bromo-1-hexene
Formule: C₆H₁₁Br
Kookpunt: 147°C
Dichtheid: 1,2169 g/cm³ (20 °C)
Vlampunt: 54°C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00000269
CAS nummer: 2695-47-8
EINECS: 220-267-9
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=567571b45e7f1e985bd23271c072cc05846765a2&vid=25be698b8d525967f2d4103d5ab351f91bada2f1&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %