trans-2-Decenal ≥93.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
D1642-25ML
D1642-5ML
TCIAD1642-25MLEA
112
EUR
TCIAD1642-25ML
TCIAD1642-5ML
trans-2-Decenal ≥93.0% (by GC)
trans-2-Decenal
Formule:
C₁₀H₁₈O Kookpunt: 80°C Vlampunt: 108°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00014679 CAS nummer: 3913-81-3 EINECS: 223-474-2 ADR: 9,III |
|
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 93.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...