1-Decene ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
D0028-25ML
D0028-500ML
TCIAD0028-25MLEA
17.6
EUR
TCIAD0028-25ML
TCIAD0028-500ML
1-Decene ≥95.0% (by GC)
1-Decene
Formule:
C₁₀H₂₀ Kookpunt: 171°C Smeltpunt: –66°C Dichtheid: 0,7408 g/cm³ (20 °C) Vlampunt: 46°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009577 CAS nummer: 872-05-9 EINECS: 212-819-2 UN: 3295 ADR: 3,III |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...