2,4,4-Trimethyl-2-pentene ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
T0780-25ML T0780-100ML
TCIAT0780-25MLEA 160 EUR
TCIAT0780-25ML TCIAT0780-100ML
2,4,4-Trimethyl-2-pentene ≥97.0% (by GC)
2,4,4-Trimethyl-2-pentene
Formule: C₈H₁₆
Kookpunt: 105°C
Dichtheid: 0,7223 g/cm³ (20 °C)
Vlampunt: 4°C
MDL Nummer: MFCD00008902
CAS nummer: 107-40-4
EINECS: 203-488-5
UN: 2050
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=954790047662eb0ecfebc277145495f3aed87366&vid=954790047662eb0ecfebc277145495f3aed87366&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %