Cyclohexylethene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

V0022-25ML V0022-5ML
TCIAV0022-25MLEA 147 EUR
TCIAV0022-25ML TCIAV0022-5ML
Cyclohexylethene ≥98.0% (by GC)
Cyclohexylethene
Formule: C₈H₁₄
Kookpunt: 128°C
Dichtheid: 1,2158 g/cm³ (18 °C)
Vlampunt: 15°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001514
CAS nummer: 695-12-5
EINECS: 211-779-3
UN: 3295
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4edd9926ce7835fbe64af6440667843db81d69b2&vid=cc22e67926a95a6c1d0294f8b48d7b21f82fbad9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR