3-Hexyne ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
H0430-25ML H0430-5ML
TCIAH0430-25MLEA 72.6 EUR
TCIAH0430-25ML TCIAH0430-5ML
3-Hexyne ≥97.0% (by GC)
3-Hexyne
Formule: C₆H₁₀
Kookpunt: 82°C
Dichtheid: 0,7231 g/cm³ (20 °C)
Vlampunt: –14°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009381
CAS nummer: 928-49-4
EINECS: 213-173-4
UN: 3295
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2c4862111b2d5eb96e10e5f26f3a8d0c1767e5e&vid=5afaf3b81dc745147416647bfa7c5425f9773d28&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %