9-Anthrealdehyde ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A0779-25G A0779-100G
TCIAA0779-25GEA 50.6 EUR
TCIAA0779-25G TCIAA0779-100G
9-Anthrealdehyde ≥99.0% (by GC)
9-Anthrealdehyde
Formule: C₁₅H₁₀O
Kookpunt: 369 °C (1013 hPa)
Smeltpunt: 101…103 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001254
CAS nummer: 642-31-9
EINECS: 211-383-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91004f85603de7461f8694c7ac3b52090e598290&vid=d442b810da78c49ace2d36eafab4807354ba3651&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Amber powder to crystal
Purity (GC) min. 99.0 %
Melting point 104.0 to 107.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR