9-Methylanthracene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M0153-1G M0153-5G
TCIAM0153-1GEA 17.6 EUR
TCIAM0153-1G TCIAM0153-5G
9-Methylanthracene ≥98.0% (by GC)
9-Methylanthracene
Formule: C₁₅H₁₂
Kookpunt: 370°C
Smeltpunt: 73…76 °C
Dichtheid: 1,06 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001261
CAS nummer: 779-02-2
EINECS: 212-299-7
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76d7837b11d2b36182b1a885bb75b7144c6c08e1&vid=cbb786bbed00ce55811eac86348a40574d137e01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 80.0 deg-C
Solubility in hot Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR