Oxamide ≥98.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

O0086-25G O0086-500G O0086-100G
TCIAO0086-25GEA 17.6 EUR
TCIAO0086-25G TCIAO0086-500G TCIAO0086-100G
Oxamide ≥98.0% (by total nitrogen basis)
Oxamide
Formule: C₂H₄N₂O₂
MW: 88.07 g/mol
MDL Nummer: MFCD00008007
CAS nummer: 471-46-5
EINECS: 207-442-5
Merck Index: 14,06918
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b25f5a3024ae2639f77f6ffa72ef8d84ba7c680&vid=9b25f5a3024ae2639f77f6ffa72ef8d84ba7c680&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Drying loss max. 1.0 %
Ignition residue (Sulfate) max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR