Benzyl cinnamate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C0358-25G
C0358-500G
TCIAC0358-25GEA
31.9
EUR
TCIAC0358-25G
TCIAC0358-500G
Benzyl cinnamate ≥98.0% (by GC)
Benzyl cinnamate
Formule:
C₁₆H₁₄O₂ |
MDL Nummer:
MFCD00004789 CAS nummer: 103-41-3 EINECS: 203-109-3 Merck Index: 14,01130 |
|
Specification Test Results
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Melting point | 35.0 to 38.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...