Broombenzeen ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B0439-25G B0439-500G
TCIAB0439-25GEA 26.4 EUR
TCIAB0439-25G TCIAB0439-500G
Broombenzeen ≥99.0% (by GC)
Broombenzeen
Formule: C₆H₅Br
Kookpunt: 156°C
Smeltpunt: –31 °C
Dichtheid: 1,495 g/cm³ (20 °C)
Vlampunt: 51°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000055
CAS nummer: 108-86-1
EINECS: 203-623-8
UN: 2514
ADR: 3,III
Merck Index: 14,01406
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=028e2d768a3b85de2dc29c775d310fc0031b4af0&vid=b8313fcf20c3130423426b8f28a483b825bd4035&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR