5-Bromosalicylaldehyde ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
5-Bromo-2-hydroxybenzaldehyde
B0893-25G
B0893-500G
TCIAB0893-25GEA
68.2
EUR
TCIAB0893-25G
TCIAB0893-500G
5-Bromosalicylaldehyde ≥98.0% (by GC, titration analysis)
5-Bromosalicylaldehyde
Formule:
C₇H₅BrO₂ Smeltpunt: 101…104 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003330 CAS nummer: 1761-61-1 EINECS: 217-167-2 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Melting point | 103.0 to 107.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...