3-Bromobenzaldehyde ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B0548-25G B0548-500G B0548-100G
TCIAB0548-25GEA 28.6 EUR
TCIAB0548-25G TCIAB0548-500G TCIAB0548-100G
3-Bromobenzaldehyde ≥98.0% (by GC)
3-Bromobenzaldehyde
Formule: C₇H₅BrO
MW: 185.02 g/mol
Kookpunt: 236°C
Vlampunt: 96°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003345
CAS nummer: 3132-99-8
EINECS: 221-526-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cde371501a7307ba955e90630b8f59ca7ace484&vid=184fd21bf7cee5b25ce71a197cd762bd41d3b701&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR