4-Bromobenzaldehyde ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B0549-25G
B0549-250G
TCIAB0549-25GEA
22
EUR
TCIAB0549-25G
TCIAB0549-250G
4-Bromobenzaldehyde ≥97.0% (by GC)
4-Bromobenzaldehyde
Formule:
C₇H₅BrO MW: 185.02 g/mol Kookpunt: 125°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003377 CAS nummer: 1122-91-4 EINECS: 214-365-0 |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 57.0 to 61.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...