1-Bromo-2-chlorobenzene ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B0569-25G
B0569-500G
TCIAB0569-25GEA
28.6
EUR
TCIAB0569-25G
TCIAB0569-500G
1-Bromo-2-chlorobenzene ≥99.0% (by GC)
1-Bromo-2-chlorobenzene
Formule:
C₆H₄BrCl Kookpunt: 204°C Dichtheid: 1,65 g/cm³ (20 °C) Vlampunt: 79°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000532 CAS nummer: 694-80-4 EINECS: 211-775-1 |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...