1-Bromo-2,6-difluorobenzeno ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D1943-25G D1943-5G
TCIAD1943-25GEA 153 EUR
TCIAD1943-25G TCIAD1943-5G
1-Bromo-2,6-difluorobenzeno ≥98.0% (by GC)
1-Bromo-2,6-difluorobenzeno
Formule: C₆H₃BrF₂
Kookpunt: 137°C
Dichtheid: 1,71 g/cm³ (20 °C)
Vlampunt: 53°C
MDL Nummer: MFCD00009894
CAS nummer: 64248-56-2
EINECS: 264-750-2
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07ff25d2650d8dd6a096cc479156a12ebd1800e3&vid=2229e47439bbad25e958dcad6b86ffe4aa4f1065&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %