Ethyl thiooxamate ≥98.0% (by GC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Thiooxamic acid ethyl ester, Ethyl amino(thioxo)acetate, Ethyl 2-amino-2-thioxoacetate

E0735-1G E0735-5G
TCIAE0735-1GEA 31.9 EUR
TCIAE0735-1G TCIAE0735-5G
Ethyl thiooxamate ≥98.0% (by GC, total nitrogen)
Ethyl thiooxamate
Formule: C₄H₇NO₂S
MW: 133.17 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00074903
CAS nummer: 16982-21-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=382ad7a3a4aa707234ef40b3f678e27627f9acd4&vid=382ad7a3a4aa707234ef40b3f678e27627f9acd4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 62.0 to 66.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR