Phenyl vinyl sulphone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Vinyl phenyl sulphone, (Vinylsulphonyl)benzene

P0982-25G P0982-5G
TCIAP0982-25GEA 153 EUR
TCIAP0982-25G TCIAP0982-5G
Phenyl vinyl sulphone ≥98.0% (by GC)
Phenyl vinyl sulphone
Formule: C₈H₈O₂S
MDL Nummer: MFCD00007554
CAS nummer: 5535-48-8
EINECS: 226-890-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 67.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR