Difenylsulfon ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Phenyl sulphone, sulphonyldibenzene
P0231-25G
P0231-500G
TCIAP0231-25GEA
17.6
EUR
TCIAP0231-25G
TCIAP0231-500G
Difenylsulfon ≥99.0% (by GC)
Difenylsulfon
Formule:
C₁₂H₁₀O₂S |
MDL Nummer:
MFCD00007548 CAS nummer: 127-63-9 EINECS: 204-853-1 Merck Index: 14,03332 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 125.0 to 130.0 deg-C |
Solubility in Acetone | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...