4-Chlorophenylhydrazinium chloride ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4-Chlorophenylhydrazine hydrochloride, 1-(4-Chlorophenyl)hydrazine hydrochloride

C0257-25G C0257-250G
TCIAC0257-25GEA 70.4 EUR
TCIAC0257-25G TCIAC0257-250G
4-Chlorophenylhydrazinium chloride ≥98.0% (by HPLC, titration analysis)
4-Chlorophenylhydrazinium chloride
Formule: C₆H₈Cl₂N₂
MW: 179.05 g/mol
MDL Nummer: MFCD00012943
CAS nummer: 1073-70-7
EINECS: 214-030-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0e704b64c88b7ab10d13d517d2a57a98e67e392&vid=b0e704b64c88b7ab10d13d517d2a57a98e67e392&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Potassium iodate Method) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR