Pentafluorophenylhydrazine ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
P0852-25G
P0852-5G
TCIAP0852-25GEA
220
EUR
TCIAP0852-25G
TCIAP0852-5G
Pentafluorophenylhydrazine ≥98.0% (by GC, titration analysis)
Pentafluorophenylhydrazine
Formule:
C₆H₃F₅N₂ MW: 198.1 g/mol Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00007574 CAS nummer: 828-73-9 EINECS: 212-586-7 |
|
Specification Test Results
Appearance | White to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 76.0 to 80.0 deg-C |
Solubility in Methanol | within almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...