3-Bromo-4-fluorobenzaldehyde ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B2143-25G B2143-5G
TCIAB2143-25GEA 110 EUR
TCIAB2143-25G TCIAB2143-5G
3-Bromo-4-fluorobenzaldehyde ≥97.0% (by GC)
3-Bromo-4-fluorobenzaldehyde
Formule: C₇H₄BrFO
Kookpunt: 312 °C (1013 hPa)
Smeltpunt: 30…32 °C
MDL Nummer: MFCD00042186
CAS nummer: 77771-02-9
EINECS: 278-764-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb88bb42e3f54b52a1458772803eaa259d1307ae&vid=84ef78ce9797fcff84a9fb4128c6c9ae225ad998&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %