1-Bromo-4-iodobenzene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4-Bromo-1-iodobenzene

B0604-25G B0604-250G
TCIAB0604-25GEA 64.9 EUR
TCIAB0604-25G TCIAB0604-250G
1-Bromo-4-iodobenzene ≥98.0% (by GC)
1-Bromo-4-iodobenzene
Formule: C₆H₄BrI
MW: 282.91 g/mol
Kookpunt: 122°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001051
CAS nummer: 589-87-7
EINECS: 209-662-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fdaeed81367ff4a506cfc83690a4ea2a517428e&vid=5c703d59891860b1c9368e72feb4fd8e9234c1c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 90.0 to 93.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR