4-Bromo-1,3-dimethoxybenzene ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-Bromo-2,4-dimethoxybenzene
B1481-25G B1481-5G
TCIAB1481-25GEA 58.3 EUR
TCIAB1481-25G TCIAB1481-5G
4-Bromo-1,3-dimethoxybenzene ≥97.0% (by GC)
4-Bromo-1,3-dimethoxybenzene
Formule: C₈H₉BrO₂
Kookpunt: 132°C
Dichtheid: 1,507 g/cm³ (20 °C)
MDL Nummer: MFCD00009844
CAS nummer: 17715-69-4
EINECS: 241-717-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b228fc464f9aad37abe4629e4436360ee7dd63d&vid=1b228fc464f9aad37abe4629e4436360ee7dd63d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow powder to lump
Purity (GC) min. 97.0 %