4-Bromophenol ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B0787-25G B0787-500G B0787-100G
TCIAB0787-25GEA 23.1 EUR
TCIAB0787-25G TCIAB0787-500G TCIAB0787-100G
4-Bromophenol ≥98.0% (by GC, titration analysis)
4-Bromophenol
Formule: C₆H₅BrO
Kookpunt: 238°C
Smeltpunt: 63 °C
Dichtheid: ~ 1,84 g/cm³ (20 °C)
Vlampunt: ≥ 110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002313
CAS nummer: 106-41-2
EINECS: 203-394-4
UN: 3077
ADR: 9,III
Merck Index: 14,01428
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c07e7e461be5aabf9bb645a748408a13a66d81ec&vid=53dd3c9344ec7e8d27e545283fa3c4178c325b01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 63.0 to 67.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR