2-Bromothiophenol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
B2347-25G B2347-5G
TCIAB2347-25GEA 135 EUR
TCIAB2347-25G TCIAB2347-5G
2-Bromothiophenol ≥98.0% (by GC)
2-Bromothiophenol
Formule: C₆H₅BrS
Kookpunt: 93°C
Dichtheid: 1,604 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004827
CAS nummer: 6320-02-1
EINECS: 228-665-4
UN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2f30371d5b047f601d794a9fa05935ddc356121&vid=0e60b6b6384ca66952d7206909c707b80f87bbea&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %