4-Bromothiophenol ≥97.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
B1385-25G B1385-5G
TCIAB1385-25GEA 67.1 EUR
TCIAB1385-25G TCIAB1385-5G
4-Bromothiophenol ≥97.0% (by GC, titration analysis)
4-Bromothiophenol
Formule: C₆H₅BrS
Kookpunt: 231°C
Smeltpunt: 70…71 °C
Dichtheid: 1,526 g/cm³ (83 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004845
CAS nummer: 106-53-6
EINECS: 203-407-3
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52e41df16c14b9503d79b71f07eaf4893ad25150&vid=67e621116cd4461d613c96b0bac0d32dc693a00b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 73.0 to 76.0 deg-C
Solubility in Methanol very faint turbidity