2-Hydroxy-m-anisaldehyde ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: o-Vanillin, 2-Hydroxy-3-methoxybenzaldehyde

H0262-25G H0262-500G H0262-100G
TCIAH0262-25GEA 29.7 EUR
TCIAH0262-25G TCIAH0262-500G TCIAH0262-100G
2-Hydroxy-m-anisaldehyde ≥99.0% (by GC)
2-Hydroxy-m-anisaldehyde
Formule: C₈H₈O₃
Kookpunt: 268°C
Smeltpunt: 40…42 °C
Dichtheid: 1,2039 g/cm³ (45 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003322
CAS nummer: 148-53-8
EINECS: 205-715-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee811599c2603ce1b901ddbceceaec385739ff96&vid=05f4aa6dd5114599b9e45d1858b8fbd629ef960f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 43.0 to 45.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR