3-Phenoxybenzaldehyde ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 3-Phenoxybenzenecarbaldehyde, m-Phenoxybenzenecarbaldehyde

P0960-25G P0960-500G
TCIAP0960-25GEA 23.1 EUR
TCIAP0960-25G TCIAP0960-500G
3-Phenoxybenzaldehyde ≥97.0% (by GC)
3-Phenoxybenzaldehyde
Formule: C₁₃H₁₀O₂
Kookpunt: 184°C
Smeltpunt: 13 °C
Dichtheid: 1,147 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003353
CAS nummer: 39515-51-0
EINECS: 254-487-1
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ffd828669eb936222351263cd9b48dc8325a2d6&vid=c4f9bc10b3c23ac4e5cde97ba222f7ec46702d9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR