Anthranilonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

A0861-25G A0861-500G A0861-100G
TCIAA0861-25GEA 23.1 EUR
TCIAA0861-25G TCIAA0861-500G TCIAA0861-100G
Anthranilonitrile ≥98.0% (by GC)
Anthranilonitrile
Formule: C₇H₆N₂
Kookpunt: 263°C
Smeltpunt: 47 °C
Dichtheid: 1,26 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007631
CAS nummer: 1885-29-6
EINECS: 217-549-9
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc44d50b7261adf5452d546348150268069cd413&vid=236fffe3d7d4814f3a4fddfb92bc138e038949a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR