3,4-Dihydroxybenzonitrile ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
D3352-25G D3352-5G
TCIAD3352-25GEA 108 EUR
TCIAD3352-25G TCIAD3352-5G
3,4-Dihydroxybenzonitrile ≥98.0% (by GC)
3,4-Dihydroxybenzonitrile
Formule: C₇H₅NO₂
Kookpunt: 311 °C (1013 hPa)
Smeltpunt: 160…163 °C
MDL Nummer: MFCD00016436
CAS nummer: 17345-61-8
EINECS: 241-367-9
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ad82479005eaae9aeb58567a0913b5498a213a6&vid=d5e7922a8216378feb8f2e2d7879c1c801baca35&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 155.0 to 159.0 deg-C