Butyl benzoate ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B0066-25ML
B0066-500ML
TCIAB0066-25MLEA
18.7
EUR
TCIAB0066-25ML
TCIAB0066-500ML
Butyl benzoate ≥99.0% (by GC)
Butyl benzoate
Formule:
C₁₁H₁₄O₂ Kookpunt: 250°C Smeltpunt: –22°C Dichtheid: 1,01 g/cm³ (20 °C) Vlampunt: 122°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009439 CAS nummer: 136-60-7 EINECS: 205-252-7 Merck Index: 14,01552 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...