Benzoic acid methyl ester ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Methyl benzoate
B0074-25ML B0074-500ML
TCIAB0074-25MLEA 19.8 EUR
TCIAB0074-25ML TCIAB0074-500ML
Benzoic acid methyl ester ≥99.0% (by GC)
Benzoic acid methyl ester
Formule: C₆H₅COOCH₃
MW: 136.15 g/mol
Kookpunt: 200°C
Smeltpunt: –12°C
Vlampunt: 91°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008421
CAS nummer: 93-58-3
EINECS: 202-259-7
Merck Index: 14,06024
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88112e38a7a258f851dd72a5e649852cbc3be3af&vid=aed1097251c094bc08b73b1e1100afee5e262304&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless clear liquid
Purity (GC) min. 99.0 %