Propyl benzoate ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B0076-25ML
B0076-500ML
TCIAB0076-25MLEA
17.6
EUR
TCIAB0076-25ML
TCIAB0076-500ML
Propyl benzoate ≥99.0% (by GC)
Propyl benzoate
Formule:
C₁₀H₁₂O₂ Kookpunt: 230°C Smeltpunt: –52 °C Dichtheid: 1,02 g/cm³ (20 °C) Vlampunt: 112°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009370 CAS nummer: 2315-68-6 EINECS: 219-020-8 |
|
Specification Test Results
Appearance | Colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...