Sodium benzoate ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Benzoic acid sodium salt
S0593-25G S0593-500G
TCIAS0593-25GEA 17.6 EUR
TCIAS0593-25G TCIAS0593-500G
Sodium benzoate ≥98.0% (by HPLC, titration analysis)
Sodium benzoate
Formule: C₆H₅COONa
MW: 144.11 g/mol
Smeltpunt: 450°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012463
CAS nummer: 532-32-1
EINECS: 208-534-8
Bereik: 01-2119460683-35
Merck Index: 14,08582
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ccdbf705cece0a6111fa02324b674e21f7723a9&vid=a7169716b3ad86c378069f610fc8d1a6d4f2ed63&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %