5-Aminosalicylic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Mesalamine, Mesalazine, 5-Amino-2-hydroxybenzoic acid, m-Aminosalicylic acid

A0317-25G A0317-500G
TCIAA0317-25GEA 25.3 EUR
TCIAA0317-25G TCIAA0317-500G
5-Aminosalicylic acid ≥98.0% (by titrimetric analysis)
5-Aminosalicylic acid
Formule: C₇H₇NO₃
Kookpunt: 347 °C (1013 hPa)
Smeltpunt: 265°C
Dichtheid: 1,568 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007877
CAS nummer: 89-57-6
EINECS: 201-919-1
Merck Index: 14,05904
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f9e57369c6da5875e497e137bdad15e670b7780&vid=c336975d3dc3845ce94088199db9aa013df94958&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light red to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in HCl almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR