Syringic acid ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4-Hydroxy-3,5-dimethoxybenzoic acid

G0014-25G G0014-500G G0014-100G
TCIAG0014-25GEA 38.5 EUR
TCIAG0014-25G TCIAG0014-500G TCIAG0014-100G
Syringic acid ≥97.0% (by titrimetric analysis)
Syringic acid
Formule: C₉H₁₀O₅
Kookpunt: 351 °C (1013 hPa)
Smeltpunt: 204…207 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002552
CAS nummer: 530-57-4
EINECS: 208-486-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b314a349fad677268f3a1b80bd5f83eb07f8e6b5&vid=aa8d126ad0bfaa5dfc44a9b5e85b43c4e8089bd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 204.0 to 212.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR