3,4-Dihydroxybenzoic acid ≥98.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
C0055-25G
C0055-100G
TCIAC0055-25GEA
47.3
EUR
TCIAC0055-25G
TCIAC0055-100G
3,4-Dihydroxybenzoic acid ≥98.0% (by HPLC)
3,4-Dihydroxybenzoic acid
Formule:
C₇H₆O₄ Kookpunt: 125 - 135 °C (0,001 -rr) Smeltpunt: 202…204 °C Dichtheid: 1,5415 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002509 CAS nummer: 99-50-3 EINECS: 202-760-0 Merck Index: 14,07894 |
|
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...