4-Phenoxybenzoic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
P1256-25G
P1256-5G
TCIAP1256-25GEA
102
EUR
TCIAP1256-25G
TCIAP1256-5G
4-Phenoxybenzoic acid ≥98.0% (by GC, titration analysis)
4-Phenoxybenzoic acid
Formule:
C₁₃H₁₀O₃ |
MDL Nummer:
MFCD00002539 CAS nummer: 2215-77-2 EINECS: 218-682-5 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 159.0 to 164.0 deg-C |
Solubility in hot Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...