1,2-Dichloorbenzeen ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: o-Dichlorobenzene
D1116-25ML D1116-500ML
TCIAD1116-25MLEA 20.9 EUR
TCIAD1116-25ML TCIAD1116-500ML
1,2-Dichloorbenzeen ≥99.0% (by GC)
1,2-Dichloorbenzeen
Formule: C₆H₄Cl₂
Kookpunt: 180°C
Smeltpunt: –17 °C
Dichtheid: 1,309 g/cm³ (20 °C)
Vlampunt: 69°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000535
CAS nummer: 95-50-1
EINECS: 202-425-9
UN: 1591
ADR: 6.1,III
Merck Index: 14,03056
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a13702c0d44de2555a3c9e4a3e3e3d7a3f5b51bc&vid=f72255d05a87cccc376fd0c84dc4b64addc8fcf3&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Specificaties
Environmentally Preferable