1,3-Dichloorbenzeen ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
D0333-25G
D0333-500G
TCIAD0333-25GEA
17.6
EUR
TCIAD0333-25G
TCIAD0333-500G
1,3-Dichloorbenzeen ≥98.0% (by GC)
1,3-Dichloorbenzeen
Formule:
C₆H₄Cl₂ Kookpunt: 172°C Smeltpunt: –25°C Dichtheid: 1,2884 g/cm³ (20 °C) Vlampunt: 62°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000573 CAS nummer: 541-73-1 EINECS: 208-792-1 UN: 3082 ADR: 9,III Merck Index: 14,03055 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...