1,4-Dichloorbenzeen ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D0687-25G D0687-500G
TCIAD0687-25GEA 17.6 EUR
TCIAD0687-25G TCIAD0687-500G
1,4-Dichloorbenzeen ≥99.0% (by GC)
1,4-Dichloorbenzeen
Formule: C₆H₄Cl₂
Kookpunt: 173,4 °C (1013 hPa)
Smeltpunt: 48…50 °C
Dichtheid: 1,458 g/cm³ (20 °C)
Vlampunt: 65 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000604
CAS nummer: 106-46-7
EINECS: 203-400-5
UN: 3077
ADR: 9,III
Merck Index: 14,03057
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1d32210e3174a3458f1bcba6fa72c808b7aaad1&vid=3e3272ffbad58c328b6a31462ae9562c47a41470&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White powder to crystal
Purity (GC) min. 99.0 %
Melting point 52.0 to 57.0 deg-C
Solubility in Methanol almost transparency