6-Chloroanthranilic acid ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
C2048-25G
C2048-5G
TCIAC2048-25GEA
80.3
EUR
TCIAC2048-25G
TCIAC2048-5G
6-Chloroanthranilic acid ≥98.0% (by HPLC, titration analysis)
6-Chloroanthranilic acid
Formule:
C₇H₆ClNO₂ Kookpunt: 330 °C (1013 hPa) Smeltpunt: 153…154 °C |
MDL Nummer:
MFCD00051530 CAS nummer: 2148-56-3 EINECS: 218-416-8 |
|
Specification Test Results
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...