4-Chloro-1,2-difluorobenzene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C1599-25G C1599-5G
TCIAC1599-25GEA 146 EUR
TCIAC1599-25G TCIAC1599-5G
4-Chloro-1,2-difluorobenzene ≥98.0% (by GC)
4-Chloro-1,2-difluorobenzene
Formule: C₆H₃ClF₂
Kookpunt: 127°C
Dichtheid: 1,35 g/cm³ (20 °C)
Vlampunt: 36°C
MDL Nummer: MFCD00042572
CAS nummer: 696-02-6
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Specificaties
Environmentally Preferable